Learn More
Echelon Biosciences Research Labs BrP-LPA 1 mg
SDP
Catalog No. 50720525
Encompass
BrP-LPA, aka LPA bromophosphonate, acts as both an Autotaxin inhibitor (94% inhibition at 10 M) and pan LPA receptor antagonist (LPA1: 1.5 M, LPA2: 1.4 M, LPA3: 1.2 M, LPA4: 0.27 M). BrP-LPA inhibits the invasiveness of NIH3T3 ras ATX cells by 40% and decreases chemotaxis by 23%. BrP-LPA reduces the size of breast (MDA-MB-231), colon (HCT-116) and melanoma (B16F10) tumors in mouse models. It also attenuates collagen-induced arthritis PPAR is not activated by BrP-LPA. This item is non returnable
This item is not returnable. View return policy
Catalog No. 50-720-525 Supplier Echelon Biosciences Research Labs Supplier No. L74161MG
BrP-LPA, aka LPA bromophosphonate, acts as both an Autotaxin inhibitor (94% inhibition at 10 M) and pan LPA receptor antagonist (LPA1: 1.5 M, LPA2: 1.4 M, LPA3: 1.2 M, LPA4: 0.27 M). BrP-LPA inhibits the invasiveness of NIH3T3 ras ATX cells by 40% and decreases chemotaxis by 23%. BrP-LPA reduces the size of breast (MDA-MB-231), colon (HCT-116) and melanoma (B16F10) tumors in mouse models. It also attenuates collagen-induced arthritis PPAR is not activated by BrP-LPA. This item is non returnable