Learn More
MSC PRO-SOURCE Back Strip Brush, Stainless Steel Back, 1 ea
Catalog No. 502817405
Encompass
This back strip brush is designed with a stainless steel back and brass bristles, providing an effective solution for various cleaning and material handling applications.
  • Bristle length of 4.0 in
  • Back strip brush width of 3/16 in
  • Overall length of 72.0 in
  • Constructed from durable brass bristle material
  • Made in the United States
Catalog No. 50-281-7405 Supplier MSC Supplier No. 09300849
This back strip brush is designed with a stainless steel back and brass bristles, providing an effective solution for various cleaning and material handling applications.
  • Bristle length of 4.0 in
  • Back strip brush width of 3/16 in
  • Overall length of 72.0 in
  • Constructed from durable brass bristle material
  • Made in the United States