Learn More
MSC Makita Chain Saw, Cordless, 40V Max, 18 in Bar, Brushless
Catalog No. 502838647
Encompass
This cordless 18 inch chain saw is designed for cutting and trimming applications, offering an alternative to gas-powered equipment. It provides zero emissions, lower noise, and reduced maintenance. Powered by a 40V max XGT battery, its brushless motor delivers power equivalent to a 42cc gas chain saw, with optimized performance under heavy loads. Features include a variable speed trigger, high chain speed, and Active Feedback-Sensing Technology.
  • Brushless motor delivers power of a 42cc gas chain saw.
  • 27% faster chain speed at 5,020 FPM.
  • Extreme Protection Technology (XPT™) engineered for improved dust and water resistance.
  • Tool-less chain adjustment for convenient operation and maintenance.
  • Adjustable automatic chain lubrication with large oil reservoir.
  • Compatible with 40V max XGT Lithium-Ion Batteries.
Catalog No. 50-283-8647 Supplier MSC Supplier No. 85354454
This cordless 18 inch chain saw is designed for cutting and trimming applications, offering an alternative to gas-powered equipment. It provides zero emissions, lower noise, and reduced maintenance. Powered by a 40V max XGT battery, its brushless motor delivers power equivalent to a 42cc gas chain saw, with optimized performance under heavy loads. Features include a variable speed trigger, high chain speed, and Active Feedback-Sensing Technology.
  • Brushless motor delivers power of a 42cc gas chain saw.
  • 27% faster chain speed at 5,020 FPM.
  • Extreme Protection Technology (XPT™) engineered for improved dust and water resistance.
  • Tool-less chain adjustment for convenient operation and maintenance.
  • Adjustable automatic chain lubrication with large oil reservoir.
  • Compatible with 40V max XGT Lithium-Ion Batteries.