Learn More
MSC Makita Chain Saw, 40V max XGT, 12 in Bar, Top Handle
Catalog No. 502838652
Encompass
This 40V XGT 12" Top Handle Chain Saw is a cordless alternative to gas-powered outdoor power equipment, offering zero emissions, lower noise, and reduced maintenance. Designed for professional arborists, it delivers up to 25% faster cutting speeds and a high power-to-weight ratio for in-tree work. Powered by a single 40V XGT battery, it features a variable speed trigger with high chain speed (0-4,882 FPM) and a brushless motor providing power equivalent to a 30cc gas chain saw.
  • Power equivalent to a 30cc gas top handle chain saw.
  • Up to 25% faster cutting speeds.
  • Runs for up to 160 cuts in 4x4 cedar with a 40V max 4.0Ah battery.
  • Cordless operation: no gas, emissions, oil mixing, or pull starts.
  • Reduced maintenance lowers operating costs.
  • IPX4 weather-resistant construction and XPT™ for durability.
Catalog No. 50-283-8652 Supplier MSC Supplier No. 88750864
This 40V XGT 12" Top Handle Chain Saw is a cordless alternative to gas-powered outdoor power equipment, offering zero emissions, lower noise, and reduced maintenance. Designed for professional arborists, it delivers up to 25% faster cutting speeds and a high power-to-weight ratio for in-tree work. Powered by a single 40V XGT battery, it features a variable speed trigger with high chain speed (0-4,882 FPM) and a brushless motor providing power equivalent to a 30cc gas chain saw.
  • Power equivalent to a 30cc gas top handle chain saw.
  • Up to 25% faster cutting speeds.
  • Runs for up to 160 cuts in 4x4 cedar with a 40V max 4.0Ah battery.
  • Cordless operation: no gas, emissions, oil mixing, or pull starts.
  • Reduced maintenance lowers operating costs.
  • IPX4 weather-resistant construction and XPT™ for durability.