Learn More
Aviva Systems Biology Corp Brsk1 Antibody - C-terminal region (ARP59832_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59832_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Brsk1. It was validated on Western Blot. . Alternate names: Gm1100, MGC99905, SAD-B, SADBApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-286-849