Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human BRSK1 (SAD1), GST Tag Recombinant Protein

Catalog No. PV4333
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PV4333 10 μg
PR8037A 100 μg
PV4344 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PV4333 Supplier Invitrogen™ Supplier No. PV4333
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Full Length.

BRSK1 (SAD1) is a serine/threonine kinase, and is a UV-induced DNA damage checkpoint kinase. BRSK1 is unbiquitously expressed, with highest levels of expression in the brain and testes. Similar to its yeast homolog, BRSK1 is thought to be involved in stress-induced cell cycle arrest. Overexpression of this protein leads to the G2/M arrest in HeLa S2 cells and UV-induced G2/M arrest could be partially abrogated by reduced expression of BRSK1 through the use of siRNA, indicating its role in DNA damage checkpoint function. More recently, it has been shown that both BRSK1 and the related protein BRSK2 are required for mammalian neuronal polarization. While BRSK1- and BRSK2-null mice were viable, double-mutant mice died within two hours of birth. Neurons from these mice showed uniformly-sized neurites as opposed to the normal long axon and multiple shorter dendrites. These neurites also displayed both axonal and dendritic markers. At least two isoforms of BRSK1 are known to exist.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q8TDC3
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50mM tris with 150mM NaCl, 0.02% Triton X-100, 2mM DTT, 50% glycerol, 0.5mM EDTA and no preservative; pH 7.5
Gene ID (Entrez) 84446
Molecular Weight (g/mol) 112.5 kDa
Name Human BRSK1 (SAD1), GST Tag
pH Range 7.5
Purification Method Purified
Quantity 10 μg
Storage Requirements -80°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias BR serine/threonine kinase 1; BR serine/threonine-protein kinase 1; Brain-selective kinase 1; brain-specific serine/threonine-protein kinase 1; Brsk1; Gm1100; hSAD1; KIAA1811; protein kinase SAD1A; RGD1563268; SAD1; SAD1 homolog; SAD1 kinase; SAD1A; SADB; SAD-B; SadB kinase short isoform; serine/threonine kinase SADB; serine/threonine kinase SAD-B; serine/threonine-protein kinase BRSK1; Serine/threonine-protein kinase SAD-B; synapses of Amphids Defective homolog 1
Common Name BRSK1
Gene Symbol Brsk1
Kinase Family CAMKL Kinase Family
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag GST-tag
Sequence Full length
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Assay Entry Biochemical binding/kinetic, Biochemical phospho-specific activity (no antibody)
Kinase Group Ser/Thr Kinases: CAMK Group
Shipping Condition Dry Ice
System Compatibility Zin-LYTE™
Target Entry BRSK1
Target Gene BR serine/threonine kinase 1
Expression System Baculovirus
Protein Family Kinases & Inhibitors
Protein Form Recombinant, Full Length
Form Liquid
Protein Subtype Serine/Threonine Kinases
Research Category Drug Discovery: Signal Transduction
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.