Learn More
CELLTREAT Scientific Products Breathable Sealing Film, Sterile, 50/CS
SDP
Catalog No. 501535150
Encompass_Preferred
CELLTREAT Scientific Products Breathable Sealing Film. Breathable Sealing Film promotes uniform gas exchange and can be pierced easily with pipet tips. Reduces outer well evaporation and cross contamination. Recommended for use between -20¿C and 120¿C. Manufactured from Rayon with Acrylic adhesive. Every case is printed with the lot number for quality traceability. Packaged sterile, 50/Re-sealable bag, 50/case.
Catalog No. 50-153-5150 Supplier CELLTREAT Scientific Products Supplier No. 229130
CELLTREAT Scientific Products Breathable Sealing Film. Breathable Sealing Film promotes uniform gas exchange and can be pierced easily with pipet tips. Reduces outer well evaporation and cross contamination. Recommended for use between -20¿C and 120¿C. Manufactured from Rayon with Acrylic adhesive. Every case is printed with the lot number for quality traceability. Packaged sterile, 50/Re-sealable bag, 50/case.