Learn More
Cayman Chemical Bruceine A, Purity: ≥98%, CAS: 25514-31-2, 10 mg
SDP
Catalog No. 502989695
Encompass_Preferred
A quassinoid with diverse biological activities; active against T. evansi (IC50 = 0.017 µg/ml); induces lipolysis in 3T3-L1 adipocytes at 160 nM; cytotoxic to HT-29, HeLa, and HL-60 cells (IC50s = 1.6, 0.6, and 0.069 µg/ml, respectively); inhibits NF-κB activity in HeLa cell extracts (IC50 = 2.2 µg/ml)
Catalog No. 50-298-9695 Supplier Cayman Chemical Supplier No. 2993610 MG
A quassinoid with diverse biological activities; active against T. evansi (IC50 = 0.017 µg/ml); induces lipolysis in 3T3-L1 adipocytes at 160 nM; cytotoxic to HT-29, HeLa, and HL-60 cells (IC50s = 1.6, 0.6, and 0.069 µg/ml, respectively); inhibits NF-κB activity in HeLa cell extracts (IC50 = 2.2 µg/ml)