Learn More
Apexbio Technology LLC Bruceine A 25514-31-2 1mg
SDP
Catalog No. 5000090737
Encompass_Preferred
Bruceine A (CAS 25514-31-2) is a natural quassinoid compound demonstrating inhibitory activity against intraerythrocytic Babesia species It is designed to suppress the proliferation of Babesia parasites thereby disrupting the parasitic lifecycle in host erythrocytes Bruceine A exerts its antiparasitic activity by inhibiting Babesia gibsoni growth In in vitro canine erythrocyte cultures Bruceine A demonstrates potent inhibition with an IC50 of approximately 7 7 nM and achieves parasite elimination at concentrations as low as 25 nM within 24 hours Based on these pharmacological properties Bruceine A holds research potential in exploring therapeutic approaches for canine babesiosis
Catalog No. 50-000-90737 Supplier Apexbio Technology LLC Supplier No. B36161
Bruceine A (CAS 25514-31-2) is a natural quassinoid compound demonstrating inhibitory activity against intraerythrocytic Babesia species It is designed to suppress the proliferation of Babesia parasites thereby disrupting the parasitic lifecycle in host erythrocytes Bruceine A exerts its antiparasitic activity by inhibiting Babesia gibsoni growth In in vitro canine erythrocyte cultures Bruceine A demonstrates potent inhibition with an IC50 of approximately 7 7 nM and achieves parasite elimination at concentrations as low as 25 nM within 24 hours Based on these pharmacological properties Bruceine A holds research potential in exploring therapeutic approaches for canine babesiosis