Learn More
Medchemexpress LLC Bruceine B | 25514-29-8 | 5 MG
SDP
Catalog No. 5000421528
Encompass_Preferred
Bruceine B inhibits protein synthesis and nucleic acid synthesis, exhibiting an IC50 of 10 nM for antimalarial activity. This product is intended for research use only and is not for patient use.
  • Inhibits protein synthesis and nucleic acid synthesis
  • Exhibits antimalarial activity with an IC50 of 10 nM
  • High purity: 99.50%
  • Solid, white to off-white appearance
  • Derived from plants Simaroubaceae Brucea javanica (L.) Merr.
  • Demonstrates cytotoxicity against various human and mouse cancer cell lines
Catalog No. 50-004-21528 Supplier Medchemexpress LLC Supplier No. HYN30135MG
Bruceine B inhibits protein synthesis and nucleic acid synthesis, exhibiting an IC50 of 10 nM for antimalarial activity. This product is intended for research use only and is not for patient use.
  • Inhibits protein synthesis and nucleic acid synthesis
  • Exhibits antimalarial activity with an IC50 of 10 nM
  • High purity: 99.50%
  • Solid, white to off-white appearance
  • Derived from plants Simaroubaceae Brucea javanica (L.) Merr.
  • Demonstrates cytotoxicity against various human and mouse cancer cell lines