Learn More
Biolog Inc BRU - Brucella Blood Agar, 1 Plate/Pkg
SDP
Catalog No. NC9647837
Encompass
This item has a minimum qty of 15 per supplier requirements.
Brucella Blood Agar (BRU) allows isolation, quantification, & identification of anaerobic bacteria from clinical specimens. This media will support the growth of aerobic & facultative bacteria if incubated appropriately. This media is prepared, stored, & dispensed under oxygen-free conditions to prevent the formation of oxidized products prior to use. BRU is also available in a 4-plate package or packaged w/ other media types in a single convenience pack.BRU agar is an enriched non-selective media that supports the growth of fastidious microorganisms. BRU agar contains casein, peptones, yeast extract, & dextrose. It is supplemented w/ vitamin K & hemin to facilitate the recovery of fastidious anaerobes, & pigment prod. of Prevotella melaninogenica. Sheep blood provides growth factors & allows the observation of hemolytic reactions such as w/ Clostridium perfringens.
Catalog No. NC9647837 Supplier Biolog Inc Supplier No. AS111
This item has a minimum qty of 15 per supplier requirements.
Brucella Blood Agar (BRU) allows isolation, quantification, & identification of anaerobic bacteria from clinical specimens. This media will support the growth of aerobic & facultative bacteria if incubated appropriately. This media is prepared, stored, & dispensed under oxygen-free conditions to prevent the formation of oxidized products prior to use. BRU is also available in a 4-plate package or packaged w/ other media types in a single convenience pack.BRU agar is an enriched non-selective media that supports the growth of fastidious microorganisms. BRU agar contains casein, peptones, yeast extract, & dextrose. It is supplemented w/ vitamin K & hemin to facilitate the recovery of fastidious anaerobes, & pigment prod. of Prevotella melaninogenica. Sheep blood provides growth factors & allows the observation of hemolytic reactions such as w/ Clostridium perfringens.