Learn More
NEOGEN Brucella Agar, 10 kg
SDP
Catalog No. 502015419
Encompass_Preferred
Brucella Agar is used for the cultivation of Brucella spp. and other fastidious microorganisms in a laboratory setting. Brucella Agar is not intended for use in the diagnosis of disease or other conditions in humans.Brucella Agar is prepared according to the APHA formula for Albimi Broth. Brucella Agar is a general-purpose medium for the cultivation of Brucella spp. and fastidious microorganisms including Streptococcus pneumoniae, Streptococcus viridans, and Neisseria meningitidis. With the addition of blood, Brucella Agar is used to determine bacterial hemolytic reactions. Brucella Agar can also be used as a base for the isolation of Campylobacter spp.For more information visit https://www.neogen.com/solutions/Microbiology/brucella-agar
Catalog No. 50-201-5419 Supplier NEOGEN Supplier No. 700003220
Brucella Agar is used for the cultivation of Brucella spp. and other fastidious microorganisms in a laboratory setting. Brucella Agar is not intended for use in the diagnosis of disease or other conditions in humans.Brucella Agar is prepared according to the APHA formula for Albimi Broth. Brucella Agar is a general-purpose medium for the cultivation of Brucella spp. and fastidious microorganisms including Streptococcus pneumoniae, Streptococcus viridans, and Neisseria meningitidis. With the addition of blood, Brucella Agar is used to determine bacterial hemolytic reactions. Brucella Agar can also be used as a base for the isolation of Campylobacter spp.For more information visit https://www.neogen.com/solutions/Microbiology/brucella-agar