Learn More
Bioworld Brucella Agar, 500 g
Catalog No. 50254285
Encompass_Preferred
Brucella AgarRecommended for the cultivation of Brucella species and other fastidious microoorganismsCan be used to determine bacterial hemolytic reactions with the addition of bloodpH: 7.0Dehydrated Appearance: light beige, homogeneous powderPrepared Appearance: yellow beige agarSoluble in boiling waterContains:Pancreatic Digest of Casein Peptic Digest of Animal Tissue Yeast Extract Sodium Chloride Dextrose Sodium Bisulfite Agar
Catalog No. 50-254-285 Supplier Bioworld Supplier No. 306200151
Brucella AgarRecommended for the cultivation of Brucella species and other fastidious microoorganismsCan be used to determine bacterial hemolytic reactions with the addition of bloodpH: 7.0Dehydrated Appearance: light beige, homogeneous powderPrepared Appearance: yellow beige agarSoluble in boiling waterContains:Pancreatic Digest of Casein Peptic Digest of Animal Tissue Yeast Extract Sodium Chloride Dextrose Sodium Bisulfite Agar