Learn More
Teknova Brucella Agar Plates with 5% Sheep's Blood. 100mm Plates, 20 Plates per Sleeve, Sterile. 4 pack.
SDP
Catalog No. 502387878
Encompass
1.00% Tryptone, 1.00% Proteose Peptone, 0.10% Dextrose, 0.20% Yeast Extract,0.50% Sodium Chloride, 0.01% Sodium Bisulfite, 1.50% Agar, 5.00% Defibrinated Sheep's Blood
Catalog No. 50-238-7878 Supplier Teknova Supplier No. B01504PK
1.00% Tryptone, 1.00% Proteose Peptone, 0.10% Dextrose, 0.20% Yeast Extract,0.50% Sodium Chloride, 0.01% Sodium Bisulfite, 1.50% Agar, 5.00% Defibrinated Sheep's Blood