Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hardy Diagnostics™ Brucella Blood Agar with Hemin and Vitamin K
SDP

Catalog No. 23001830 Shop All Hardy Diagnostics Products
Change view
Click to view available options
15 x 100 mm Plate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
23-001-830 15 x 100 mm Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-001-830 Supplier Hardy Diagnostics™ Supplier No. A30
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Brucella Agar with Hemin and Vitamin K for the cultivation of anaerobes.

  • A non-inhibitory general growth medium for all Gram-positive and Gram-negative anaerobic bacteria.
  • Optimized formula for enhanced growth characteristics of anaerobes.
  • Contains 5% sheep blood from a veterinary supervised, controlled herd of donor animals.
  • Blood is guaranteed to be from healthy antibiotic-free animals.
  • Optically clear polystyrene for distortion-free viewing.
  • Stacking rings guard against slippage when moving stacked plates.
  • Packaged in breathable sleeves of 10 plates in a breathable bag that prevents buildup of condensation and excess moisture.

Specifications

Manufacturing Origin United States
Media Type Brucella Blood Agar with Hemin and Vitamin K
Product Type Agar Plate
Sterility Non-sterile
Target Organism Class Anaerobes
Test Type Microbial Detection
Packaging Type Plate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.