Learn More
Biolog Inc BRU BROTH - Brucella Broth, 4mL, 10 Tubes/Pkg
SDP
Catalog No. NC9988021
Encompass
This item has a minimum qty of 2 per supplier requirements.
Brucella Broth (BRU BROTH) is an enriched non-selective media intended for the cultivation of most anaerobic bacteria and other fastidious microorganisms. This media will support the growth of aerobic and microaerophilic bacteria if incubated appropriately. This media is prepared, stored, and dispensed under oxygen-free conditions to prevent the formation of oxidized products prior to use.BRU BROTH is an enriched non-selective broth media. The basal media consists of casein and peptones to facilitate the growth of a wide variety of microorganisms. Yeast extract is provided as a source of trace vitamins and micronutrients. A small volume of dextrose is added as a carbohydrate source, to prevent acid build up. This media is supplemented with vitamin K1 and hemin to facilitate the recovery and growth of Prevotella melaninogenica.
Catalog No. NC9988021 Supplier Biolog Inc Supplier No. AS105
This item has a minimum qty of 2 per supplier requirements.
Brucella Broth (BRU BROTH) is an enriched non-selective media intended for the cultivation of most anaerobic bacteria and other fastidious microorganisms. This media will support the growth of aerobic and microaerophilic bacteria if incubated appropriately. This media is prepared, stored, and dispensed under oxygen-free conditions to prevent the formation of oxidized products prior to use.BRU BROTH is an enriched non-selective broth media. The basal media consists of casein and peptones to facilitate the growth of a wide variety of microorganisms. Yeast extract is provided as a source of trace vitamins and micronutrients. A small volume of dextrose is added as a carbohydrate source, to prevent acid build up. This media is supplemented with vitamin K1 and hemin to facilitate the recovery and growth of Prevotella melaninogenica.