Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Research Products International Corp Brucella Broth, 500 grams, Dehydrated Powder
SDP

Catalog No. 501368124
Encompass_Preferred

Brucella Broth is used for cultivation, enrichment and isolation of Brucella species and other fastidious organisms. Supplied as dehydrated powder.

Ingredients:

Pancreatic Digest of Casein: 10 g/L
Peptic Digest, Animal Tissue: 10 g/L
Yeast Extract: 2 g/L
Sodium Chloride: 5 g/L
Dextrose: 1 g/L
Sodium Bisulfite: 0.01 g/L
Final pH: 7±0.1 at 25°C

Catalog No. 50-136-8124 Supplier Research Products International Corp Supplier No. B62000500.0
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Brucella Broth is used for cultivation, enrichment and isolation of Brucella species and other fastidious organisms. Supplied as dehydrated powder.

Ingredients:

Pancreatic Digest of Casein: 10 g/L
Peptic Digest, Animal Tissue: 10 g/L
Yeast Extract: 2 g/L
Sodium Chloride: 5 g/L
Dextrose: 1 g/L
Sodium Bisulfite: 0.01 g/L
Final pH: 7±0.1 at 25°C

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.