Learn More
Bioworld Brucella Broth, 1 kg
Supplier:  Bioworld 306200162
Encompass_Preferred
Brucella brothRecommended for the cultivation of Brucella species and other fastidious microoorganismspH: 7.0Dehydrated Appearance: light to medium beige, homogeneous powderPrepared Appearance: gold to beige brothSoluble in waterContains:Pancreatic Digest of Casein Peptic Digest of Animal Tissue Yeast Extract Sodium Chloride Dextrose Sodium Bisulfite
Catalog No. 50-254-289