Learn More
ATCC Mycoplasma felis
Catalog No. 502381517
Encompass
This strain of Mycoplasma felis was isolated from the eye of a cat with conjunctivitis. This culture is a bacterial type strain and is propagated in a 5% CO2 atmosphere on Mycoplasma Medium.
Catalog No. 50-238-1517 Supplier ATCC Supplier No. 23391
This strain of Mycoplasma felis was isolated from the eye of a cat with conjunctivitis. This culture is a bacterial type strain and is propagated in a 5% CO2 atmosphere on Mycoplasma Medium.