Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Brucine Sulfate Heptahydrate

Catalog No. 23786825G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-786-825G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-786-825G Supplier Sigma Aldrich Supplier No. 23786825G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Brucine Sulfate Heptahydrate

Specifications

CAS 23279-22-3
Melting Point 158°C to 160°C
Molecular Formula C9H12N2O3 -+ 2HCl -+ H2O
Linear Formula H2NC6H3-4-(OH)CH2CH(NH2)CO2H -+ 2HCl -+ H2O
MDL Number MFCD00150105
Quantity 5 g
Molecular Weight (g/mol) 287.14 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.