Learn More
Selleck Chemical LLC Brucine sulfate heptahydrate
SDP
Supplier:  Selleck Chemical LLC S61721G
Encompass_Preferred
Brucine is an alkaloid that acts as an antagonist at glycine receptors and paralyzes inhibitory neurons
Catalog No. 50-000-03170