Learn More
Selleck Chemical LLC Brucine sulfate heptahydrate
SDP
Catalog No. 5000003692
Encompass_Preferred
Brucine is an alkaloid that acts as an antagonist at glycine receptors and paralyzes inhibitory neurons
Catalog No. 50-000-03692 Supplier Selleck Chemical LLC Supplier No. S617225MG
Brucine is an alkaloid that acts as an antagonist at glycine receptors and paralyzes inhibitory neurons