Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brucine Sulfate-Sulfuric Acid Solution, Reagents

Catalog No. 01335743
Change view
Click to view available options
Quantity: 1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01-335-743 Quantity: 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-335-743 Supplier Reagents Holdings Llc Supplier No. CB1460001C
Only null left
Add to Cart
Add to Cart

Brucine Sulfate-Sulfuric Acid Solution, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Brucine Sulfate-Sulfuric Acid Solution
CAS 7664-93-9, 7732-18-5, 7732-18-5
Color Colorless to Light Yellow
Packaging Glass Bottle
Quantity 1 L
Specific Gravity 1.58
Grade Reagent
Physical Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.