Learn More
Bruker Daltonics BRUKER HCCA PORTION 10 TUBE
Catalog No. NC0251092
Encompass_Preferred
BRUKER HCCA PORTION 10 TUBE. Store in the dark at temperatures below 0C. Stability of vacuum-dried HCCA at least 2 years.
Catalog No. NC0251092 Supplier Bruker Daltonics Supplier No. 8255344
BRUKER HCCA PORTION 10 TUBE. Store in the dark at temperatures below 0C. Stability of vacuum-dried HCCA at least 2 years.