Description
- True benchtop solutions requiring less than 3 feet of counter space
- FDA cleared library with 424 organisms in 333 groups covering anaerobes, Gram-positive bacteria, Gram negative bacteria, and yeast
- Low-noise operating system
- 110 V electrical requirements, with minimal heat output
- Automated workflow enhancement and prep instruments available
- Real time microbiology – no batching required with BD Bruker MALDI Biotyper workflow
- Bruker Bacterial Test Standard (BTS) simplifies QC – easier and more convenient than other commercially available strains
- Disposable target plates provide 96 spots and a unique barcode for full traceability, with no time consuming cleanup
- The BD EpiCenter Data Management System's Plate Mapping module allows users to spot samples on MALDI target plates while simultaneously connecting them to appropriate BD Phoenix susceptibility panels
- Straightforward result interpretation
Specifications
Specifications
| Description | Mass Spectrometry Instrument |
| Type | Bruker MALDI Biotyper Sirius System |