Description
Experience the benefits of the Bruker MALDI Biotyper
- Rapid identification of organisms provides critical information to clinicians
- Cost-effective, highly accurate system is applicable to a wide range of microorganisms
- System provides a seamless Lean workflow with the DxM MicroScan WalkAway System and MicroScan WalkAway plus
Make every minute count
- Gold-standard MALDI-TOF technology provides rapid identification of isolates
- System supports antibiotic stewardship by allowing physicians to better manage treatment
- System provides streamlined Lean workflow for maximum productivity
- High throughput enables faster TATs
Get flexible options on demand
- Single, open, comprehensive database saves time and streamlines workflow
- Target plates are available in a variety of sizes, ranging from 24 to 96 wells. Options include disposable, reusable and BigAnchorChip™ target plates for liquid samples
- Small-footprint system fits easily into the most space-constrained laboratories with a compact, ergonomic and environmentally friendly design
Specifications
Specifications
| Type | Clinical Chemistry Analyzer |
| For Use With (Equipment) | Beckman Coulter Analyzers |