Learn More
Sigma Aldrich Fine Chemicals Biosciences Bruker standard solvent 100ML, CAS Number 76-05-1 , MDL number MFCD00004169
Catalog No. NC1465326
Encompass
Bruker standard solvent for MALDI MS, 50% ACN + 47.5% H2O + 2.5% TFA . CAS Number 76-05-1 Linear Formula CF3COOH Molecular Weight 114.02 MDL number MFCD00004169. 100ML. Package: Glass Bottle
Catalog No. NC1465326 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 900666100ML
Bruker standard solvent for MALDI MS, 50% ACN + 47.5% H2O + 2.5% TFA . CAS Number 76-05-1 Linear Formula CF3COOH Molecular Weight 114.02 MDL number MFCD00004169. 100ML. Package: Glass Bottle