Learn More
Sigma Aldrich Fine Chemicals Biosciences BrukerR SampleJet NMR tube
Catalog No. 502900395
Encompass
Wilmad NMR tubes with authentic Bruker(R) SampleJet(TM) caps without code up to 600 MHz. Do not use with ceramic spinners.
Catalog No. 50-290-0395 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. WIMWG30004SJ100EA
Wilmad NMR tubes with authentic Bruker(R) SampleJet(TM) caps without code up to 600 MHz. Do not use with ceramic spinners.