Learn More
Aviva Systems Biology Corp BRUNOL6 Antibody - middle region (ARP41078_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41078_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against BRUNOL6. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human BRUNOL6. Alternate names: CELF6, BRUNOL6ApplicationWB. Species reactivity: Cow, Guinea Pig, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-57712