Learn More
Grainger Brush 2.5 Flat Sash PET/Nylon 2 15/16 L
Catalog No. 502752883
Encompass_Preferred
The Grainger 14C524 is a Westward brand 1/4-inch drive socket set. It contains both standard and deep sockets ranging in size from 4mm to 14mm. The set includes a total of 15 chrome vanadium sockets stored in a molded plastic case. The sockets have a satin finish and feature a 6-point design for gripping fasteners. This socket set is ideal for automotive and machinery applications that require metric socket sizes. The case keeps the sockets organized and protected.
Catalog No. 50-275-2883 Supplier Grainger Supplier No. 14C524
The Grainger 14C524 is a Westward brand 1/4-inch drive socket set. It contains both standard and deep sockets ranging in size from 4mm to 14mm. The set includes a total of 15 chrome vanadium sockets stored in a molded plastic case. The sockets have a satin finish and feature a 6-point design for gripping fasteners. This socket set is ideal for automotive and machinery applications that require metric socket sizes. The case keeps the sockets organized and protected.