Learn More
Grainger Brush 2 Angle Sash China Hair 2 5/8 L
Catalog No. 502752325
Encompass_Preferred
The Grainger 14C525 is a 14-inch diameter, 525 CFM, direct drive exhaust fan. It features a thermally protected PSC motor and is designed for general ventilation applications. The fan has a galvanized steel housing with baked enamel finish and includes adjustable motor mounts. It requires 115V power and is ETL listed for safety. The 14C525 can provide powerful airflow for industrial and commercial ventilation needs. With a 1/3 HP motor, it offers an optimal balance of airflow and energy efficiency. The adjustable mounting and corrosion-resistant housing make this a durable and versatile exhaust fan option.
Catalog No. 50-275-2325 Supplier Grainger Supplier No. 14C525
The Grainger 14C525 is a 14-inch diameter, 525 CFM, direct drive exhaust fan. It features a thermally protected PSC motor and is designed for general ventilation applications. The fan has a galvanized steel housing with baked enamel finish and includes adjustable motor mounts. It requires 115V power and is ETL listed for safety. The 14C525 can provide powerful airflow for industrial and commercial ventilation needs. With a 1/3 HP motor, it offers an optimal balance of airflow and energy efficiency. The adjustable mounting and corrosion-resistant housing make this a durable and versatile exhaust fan option.