Learn More
Research Products International Corp Brush, Burette, White Nylon Bristle, 36 Inch, 12 per Case
SDP
Catalog No. 501367780
Encompass
Long flexible handles reach into burettes, pipets and long tubing for cleaning. All brushes have a fan tip to prevent scratching.
Catalog No. 50-136-7780 Supplier Research Products International Corp Supplier No. 610292
Long flexible handles reach into burettes, pipets and long tubing for cleaning. All brushes have a fan tip to prevent scratching.