Learn More
Hach Company Brush, Camel´s Hair
Supplier:  Hach Company 2103200
Encompass
For cleaning delicate instruments or to ensure complete transfer of samples when weighing. Soft, brown hair is cemented into a metal holder. Hairs are cut to the same length—14 mm (0.5"). Can be attached to Micro Spatula, Prod. No. 1225600.
Catalog No. 50-199-5387