Learn More
Med Vet International Sterile Surgical Scrub Brush Dry Sterile, 30/Box
Catalog No. 501180275
Encompass_Preferred
Soft outer bristles clean thoroughly without skin irritation. Inner bristles are shorter and contoured for effective deep cleaning of fingers, nails and cuticles. Brush flexes for easier scrubbing between and around fingers. Convenient peel-open package.
Catalog No. 50-118-0275 Supplier Med Vet International Supplier No. BR125X30
Soft outer bristles clean thoroughly without skin irritation. Inner bristles are shorter and contoured for effective deep cleaning of fingers, nails and cuticles. Brush flexes for easier scrubbing between and around fingers. Convenient peel-open package.