Learn More
Gilson Company Inc BRUSH FIN GILSON TSA170
Catalog No. NC9359843
Encompass_Preferred
Fine Sieve Brush , soft, black bristle, 1-1/4 in. diameter, 5-3/4 in. long, It has a nickeled steel ferrule, clear lacquered wood handle and is ideal for cleaning No. 16 and finer round sieves and pans
Catalog No. NC9359843 Supplier Gilson Company Inc Supplier No. TSA170
Fine Sieve Brush , soft, black bristle, 1-1/4 in. diameter, 5-3/4 in. long, It has a nickeled steel ferrule, clear lacquered wood handle and is ideal for cleaning No. 16 and finer round sieves and pans