Learn More
Chemglass Life Sciences Flexible Brush, Glassware Cleaning, 48" OAL
SDP
Catalog No. 501310995
Encompass_Preferred
Flexible Brush, Glassware Cleaning, 48" OAL. Brushes have polypropylene bristles and a flexible galvanized steel wiring. Fits tubes with an I.D. of 1/2¿ to 1¿. Excellent for cleaning long tubular items. MADE IN USA.
Catalog No. 50-131-0995 Supplier Chemglass Life Sciences Supplier No. CG114202
Flexible Brush, Glassware Cleaning, 48" OAL. Brushes have polypropylene bristles and a flexible galvanized steel wiring. Fits tubes with an I.D. of 1/2¿ to 1¿. Excellent for cleaning long tubular items. MADE IN USA.