Learn More
Chemglass Life Sciences Brush, Glassware Cleaning, Foam Tip, 13" OAL
SDP
Supplier:  Chemglass Life Sciences CG114401
Encompass_Preferred
Brush, Glassware Cleaning, Foam Tip, 13" OAL. These laboratory brushes feature polypropylene bristles with foam bristles at the tip. Excellent for use with wide mouth bottles, graduated cylinders, beakers etc. Width of brush tip is 2¿. MADE IN USA.
Catalog No. 50-131-0997