Learn More
Research Products International Corp Brush, Nessler, White Bristle, 19 Inches, 12 per Case
SDP
Catalog No. 501367578
Encompass
Taller and more narrow than regular test tubes. High Form Nessler tubes require a brush that will easily fit thru the cap opening and provide extra cleaning action in the corners and bottom of tubes.
Catalog No. 50-136-7578 Supplier Research Products International Corp Supplier No. 610326
Taller and more narrow than regular test tubes. High Form Nessler tubes require a brush that will easily fit thru the cap opening and provide extra cleaning action in the corners and bottom of tubes.