Learn More
Grainger Brush On Pipe and Gasket Joint 16 oz
Catalog No. 502764715
Encompass_Preferred
The Grainger 170H28 is a 28-inch, high-visibility, Class 2 safety vest made of polyester mesh with 2-inch reflective stripes. It features a zipper front closure, an ANSI 107-2015 Type R compliant design, and comes in orange or lime color options. This lightweight, breathable vest is intended to provide visibility and meet safety requirements for roadway, utility, and construction work zones.
Catalog No. 50-276-4715 Supplier Grainger Supplier No. 170H28
The Grainger 170H28 is a 28-inch, high-visibility, Class 2 safety vest made of polyester mesh with 2-inch reflective stripes. It features a zipper front closure, an ANSI 107-2015 Type R compliant design, and comes in orange or lime color options. This lightweight, breathable vest is intended to provide visibility and meet safety requirements for roadway, utility, and construction work zones.