Learn More
Med Vet International Bd E-Z Scrub PreoperatIVe Surgical Scrub Brush Impregnated With 4% Chg (Red), 30/Box
Catalog No. 502830440
Encompass_Preferred
For over three decades, the BD E-Z Scrub™ brand has been considered the "gold standard" in single-use disposable scrubs. BD E-Z Scrub™ products offer a wide range of efficacious antimicrobial formulations specifically designed to enhance performance while reducing irritation to the skin. Proprietary polyethylene soft bristles enhance mechanical action while reducing skin irritation. Soft sponge provides gentle cleaning on irritated areas or when firm mechanical action is not required. Ergonomic brush design allows greater user comfort and effectiveness, ultimately improving compliance.
Catalog No. 50-283-0440 Supplier Med Vet International Supplier No. BD371073Z
For over three decades, the BD E-Z Scrub™ brand has been considered the "gold standard" in single-use disposable scrubs. BD E-Z Scrub™ products offer a wide range of efficacious antimicrobial formulations specifically designed to enhance performance while reducing irritation to the skin. Proprietary polyethylene soft bristles enhance mechanical action while reducing skin irritation. Soft sponge provides gentle cleaning on irritated areas or when firm mechanical action is not required. Ergonomic brush design allows greater user comfort and effectiveness, ultimately improving compliance.