Learn More
Ted Pella Inc Sable Brushes, #0, 1.3mm W x 8.0mm L, 3/pk
SDP
Catalog No. NC9691026
Encompass
For precise manipulation of TEM specimen grids and samples, dusting of SEM samples and photographic materials or photographic spotting. Sable hairs are springy and relatively fine. Length of hairs are short for better manipulation. Enameled wood handles and seamless nickel ferrules. Offered in packages of 3 brushes. Measurements taken just above ferrule.
Catalog No. NC9691026 Supplier Ted Pella Inc Supplier No. 11810
For precise manipulation of TEM specimen grids and samples, dusting of SEM samples and photographic materials or photographic spotting. Sable hairs are springy and relatively fine. Length of hairs are short for better manipulation. Enameled wood handles and seamless nickel ferrules. Offered in packages of 3 brushes. Measurements taken just above ferrule.