Learn More
Electron Microscopy Sciences Brush with Fiberglass Ferrules
SDP
Catalog No. 501926517
Encompass_Preferred
These scratch brushes with pen shaped handles are used for fine deburring, polishing, as well as rust removal and the cleaning and removal of deposited wax. They are offered with metal ferrules for improved durability. The overall length is 4 1/4" long, and the brush is 1/8" wide.
Material: Fiberglass Ferrules
Catalog No. 50-192-6517 Supplier Electron Microscopy Sciences Supplier No. 6558205
These scratch brushes with pen shaped handles are used for fine deburring, polishing, as well as rust removal and the cleaning and removal of deposited wax. They are offered with metal ferrules for improved durability. The overall length is 4 1/4" long, and the brush is 1/8" wide.
Material: Fiberglass Ferrules