Learn More
MSC Rust-Oleum Brush-Wash Cleaner & Renewer, 32 oz, Biodegradable, Non-Toxic
Catalog No. 502837856
Encompass
This 32 oz solution is designed for cleaning and renewing brushes. It is an eco-friendly, non-toxic, and water-based formula that is biodegradable and non-flammable. It effectively cleans and conditions brushes, providing a safe cleaning experience without toxic vapors.
  • Safely cleans and reconditions synthetic bristle brushes and roller covers
  • Biodegradable and non-toxic formula
  • Safe alternative to solvent-based brush cleaners
  • EPA Safer Choice Certified
  • Effectively removes paint, oil, varnish, lacquer, enamel, and shellac
  • Features a refreshing citrus scent
  • Super concentrated, ready-to-use formula
  • Do not use on natural bristle brushes
Catalog No. 50-283-7856 Supplier MSC Supplier No. 36156669
This 32 oz solution is designed for cleaning and renewing brushes. It is an eco-friendly, non-toxic, and water-based formula that is biodegradable and non-flammable. It effectively cleans and conditions brushes, providing a safe cleaning experience without toxic vapors.
  • Safely cleans and reconditions synthetic bristle brushes and roller covers
  • Biodegradable and non-toxic formula
  • Safe alternative to solvent-based brush cleaners
  • EPA Safer Choice Certified
  • Effectively removes paint, oil, varnish, lacquer, enamel, and shellac
  • Features a refreshing citrus scent
  • Super concentrated, ready-to-use formula
  • Do not use on natural bristle brushes