Learn More
HITOBIOTEC BRUSHES SET 112, 7 PCS
Catalog No. NC1514907
Encompass
Brushes set for microtome / cryostat cleaning and histology section manipulation. It is designed to be used for dusting microtome / cryostat and laying paraffin section in water bath, transfer free floating section into buffer, mounting paraffin, frozen and free floating section on slide.Tip No. 0, 2, 4, 6, 8, 10, 127 pcs/pack
Catalog No. NC1514907 Supplier HITOBIOTEC Supplier No. HTHS0112
Brushes set for microtome / cryostat cleaning and histology section manipulation. It is designed to be used for dusting microtome / cryostat and laying paraffin section in water bath, transfer free floating section into buffer, mounting paraffin, frozen and free floating section on slide.Tip No. 0, 2, 4, 6, 8, 10, 127 pcs/pack