Learn More
Chemglass Life Sciences Brushless DC Motor Only, 3/8HP, 24V, 3/4” Shaft dia., 30-500 RPM, 43in-lb
SDP
Catalog No. 5000980694
Encompass_Preferred
Features:compact Overall Height Brushless DC Motor. Mini USB port for Control or Data Logging of RPM/Torque trends. Torque Compensation – Maintains set RPM with Viscosity Changes. Chemstir 24 volt Brushless DC motor features high torque, compact size, and no maintenance. Brushless gear-motor is for use with 10L thru 100L process reactors. Must be used in conjunction with a CG-2033-V-75 controller. The motor faceplate bolt pattern mounts directly to Chemglass CG-1965-X and CG-1968-X style reactor support frames. CG-2033-V Digital Motor Controller Chemglass / 115v/230v. TFT 3. 5” Display for All Setting Information at a GlanceWi-Fi Connection toErmes Platform to monitor status and data logging on the cloudTimer For Unattended OperationsReal Time Torque and Speed Graph MonitoringIntuitive Knob To Navigate The Menu And Start Stop OperationsCompatible with CG-2033-B 3/8 HP Brushless MotorUSB interface for use with PC software to data log, monitor, Turn off/ Turn on. USB also allow Software
Catalog No. 50-009-80694 Supplier Chemglass Life Sciences Supplier No. CG2033B38
Features:compact Overall Height Brushless DC Motor. Mini USB port for Control or Data Logging of RPM/Torque trends. Torque Compensation – Maintains set RPM with Viscosity Changes. Chemstir 24 volt Brushless DC motor features high torque, compact size, and no maintenance. Brushless gear-motor is for use with 10L thru 100L process reactors. Must be used in conjunction with a CG-2033-V-75 controller. The motor faceplate bolt pattern mounts directly to Chemglass CG-1965-X and CG-1968-X style reactor support frames. CG-2033-V Digital Motor Controller Chemglass / 115v/230v. TFT 3. 5” Display for All Setting Information at a GlanceWi-Fi Connection toErmes Platform to monitor status and data logging on the cloudTimer For Unattended OperationsReal Time Torque and Speed Graph MonitoringIntuitive Knob To Navigate The Menu And Start Stop OperationsCompatible with CG-2033-B 3/8 HP Brushless MotorUSB interface for use with PC software to data log, monitor, Turn off/ Turn on. USB also allow Software