Learn More
Bellco Glass, Inc. Brushless Digital Roller Drum 0.5-15 rpm 230V
SDP
Catalog No. 501946095
Encompass_Preferred
The Bellco Brushless Digital Roller Drum Drive is an all purpose roller drum for blood, tissue culture, cells and virus applications. The standard speed ranges are 0.5 to 15 and 2 to 60 RPM with other speed ranges available as custom items. Four drums are available with capacity of 164 or 351 tubes of 16 mm diameters, 156 tubes of 25 mm diameter or 128 tubes for most 50 ml conical bottom centrifuge tubes. (Drum sold separately). MADE IN THE USA
Catalog No. 50-194-6095 Supplier Bellco Glass, Inc. Supplier No. 773641230
The Bellco Brushless Digital Roller Drum Drive is an all purpose roller drum for blood, tissue culture, cells and virus applications. The standard speed ranges are 0.5 to 15 and 2 to 60 RPM with other speed ranges available as custom items. Four drums are available with capacity of 164 or 351 tubes of 16 mm diameters, 156 tubes of 25 mm diameter or 128 tubes for most 50 ml conical bottom centrifuge tubes. (Drum sold separately). MADE IN THE USA