Learn More
Bellco Glass, Inc. Brushless Digital Roller Drum Low Profile 0.5-15 rpm 115V
SDP
Catalog No. 501946098
Encompass_Preferred
The Bellco Low Profile Brushless Digital Roller Drum Drive is an all purpose roller drum for blood, tissue culture, cells and virus applications. The standard speed ranges are 0.5 to 15 and 2 to 60 RPM with other speed ranges available as custom items. Two drums are available with capacity of 106 tubes of 18 mm diameters, or 60 tubes of 25 mm diameter. (Drum & tubes sold separately). MADE IN THE USA
Catalog No. 50-194-6098 Supplier Bellco Glass, Inc. Supplier No. 773661115
The Bellco Low Profile Brushless Digital Roller Drum Drive is an all purpose roller drum for blood, tissue culture, cells and virus applications. The standard speed ranges are 0.5 to 15 and 2 to 60 RPM with other speed ranges available as custom items. Two drums are available with capacity of 106 tubes of 18 mm diameters, or 60 tubes of 25 mm diameter. (Drum & tubes sold separately). MADE IN THE USA