Learn More
AVANTOR MASTERFLEX (VWR LLC) BRUSHLESS MASTERFLEX PUMP
Catalog No. NC9530353
Encompass
  • Designed for laboratory and industrial fluid transfer applications.
  • Features a brushless motor for reliable performance.
  • Offers flow rates from 0.4 to 8.0 L/min and 40 ml/min to 8000 ml/min.
  • Robust enclosure with IP55 rating provides high durability.
  • Compact and lightweight design allows for easy portability.
  • Variable-speed digital control ensures precise speed adjustment.
  • Integrated LED display shows percent speed with 1% resolution.
  • Supports a maximum RPM range of 33 to 650 rpm.
  • Operates within a voltage range of 90 to 260 VAC.
  • Includes three rollers for enhanced fluid handling capabilities.
  • Single channel operation for simplified use.
Catalog No. NC9530353 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX7796310
Add to Cart
Add to Cart
Request a Quote
  • Designed for laboratory and industrial fluid transfer applications.
  • Features a brushless motor for reliable performance.
  • Offers flow rates from 0.4 to 8.0 L/min and 40 ml/min to 8000 ml/min.
  • Robust enclosure with IP55 rating provides high durability.
  • Compact and lightweight design allows for easy portability.
  • Variable-speed digital control ensures precise speed adjustment.
  • Integrated LED display shows percent speed with 1% resolution.
  • Supports a maximum RPM range of 33 to 650 rpm.
  • Operates within a voltage range of 90 to 260 VAC.
  • Includes three rollers for enhanced fluid handling capabilities.
  • Single channel operation for simplified use.