Learn More
Chemglass Life Sciences Brushless DC Motor Only, 1/11 HP, 24V, 35-417 RPM, 10in-lb, w/Adapter Plate, Holding Rod and Keyless Chuck
SDP
Catalog No. 501531404
Encompass_Preferred
Brushless DC Motor Only, 1/11 HP, 417RPM, w/Adapter Plate, Holding Rod and Keyless Chuck. Chemstir 24 volt Brushless DC motor features high torque, compact size, and no maintenance. Brushless gear-motor is for use with 250mL thru 5L benchtop process reactors. Must be used in conjunction with a CG-2033-B-55 digital display controller. Support plate and rod design allows the motor to be mounted on the ChemRxnHub stand or on lattice in most hoods. Supplied complete with a Nylon body keyless chuck and a CG-9252-01 for mounting on lattice in hood area.
Catalog No. 50-153-1404 Supplier Chemglass Life Sciences Supplier No. CG2033B11
Brushless DC Motor Only, 1/11 HP, 417RPM, w/Adapter Plate, Holding Rod and Keyless Chuck. Chemstir 24 volt Brushless DC motor features high torque, compact size, and no maintenance. Brushless gear-motor is for use with 250mL thru 5L benchtop process reactors. Must be used in conjunction with a CG-2033-B-55 digital display controller. Support plate and rod design allows the motor to be mounted on the ChemRxnHub stand or on lattice in most hoods. Supplied complete with a Nylon body keyless chuck and a CG-9252-01 for mounting on lattice in hood area.