Learn More
MSC Rubbermaid Container, 32 gal, Round, Polyethylene, Gray
Supplier: MSC 77671915
This durable, heavy-duty container is designed for efficient waste management in commercial environments. Constructed with high-quality polyethylene and a UV inhibitor, it resists fading, warping, cracking, or crushing for long-lasting use.
- Venting channels make liner removal up to 50% easier.
- Integrated cinches secure liners for efficient, knot-free changes.
- Reinforced handles resist tearing and damage from heavy loads.
- Reinforced base withstands dragging over rough surfaces.
- Contoured base handles improve grip and ergonomics.
- Integrated rib structure in the rim increases strength and resists crushing.
- Meets NSF 2, 21 approval and USDA Meat and Poultry Group listed.
- Stackable design enables nesting for easy transport and storage.
- Seamless construction simplifies cleaning.